Assay Details
| Gene Symbol: mideasa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045193.1 | NP_001038658.1 | 11 - 12 | 3218 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mideasa| Gene Symbol: mideasa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045193.1 | NP_001038658.1 | 11 - 12 | 3218 | 71 | |
Set Membership: |
|