Assay Details
| Gene Symbol: mideasa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045193.1 | NP_001038658.1 | 6 - 7 | 2709 | 91 | |
| XM_005160589.4 | XP_005160646.1 | 6 - 7 | 2896 | 91 | ||
More Information
Set Membership: |
|

mideasa
Genomic Map