Assay Details
| Gene Symbol: cluha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077560.1 | NP_001071028.1 | 11 - 12 | 2582 | 78 | |
| XM_005157587.4 | XP_005157644.1 | 11 - 12 | 2385 | 78 | ||
| XM_021481439.1 | XP_021337114.1 | 11 - 12 | 2629 | 78 | ||
| GenBank mRNA | BC125829.1 | - | 11 - 12 | 2582 | 78 | |
More Information
Set Membership: |
|

cluha
Genomic Map