Assay Details
| Gene Symbol: st14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077737.1 | NP_001071205.1 | 14 - 15 | 1936 | 81 | |
| XM_005161204.4 | XP_005161261.1 | 17 - 18 | 2225 | 81 | ||
| XM_009295332.3 | XP_009293607.1 | 17 - 18 | 2145 | 81 | ||
| GenBank mRNA | BC125837.1 | - | 14 - 15 | 1936 | 81 | |
| BC165661.1 | - | 14 - 15 | 1723 | 81 | ||
More Information
Set Membership: |
|

st14b
Genomic Map