Assay Details
| Gene Symbol: st14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077737.1 | NP_001071205.1 | 3 - 4 | 630 | 86 | |
| XM_005161204.4 | XP_005161261.1 | 4 - 5 | 586 | 86 | ||
| XM_009295332.3 | XP_009293607.1 | 4 - 5 | 506 | 86 | ||
| GenBank mRNA | BC125837.1 | - | 3 - 4 | 630 | 86 | |
| BC165661.1 | - | 3 - 4 | 417 | 86 | ||
More Information
Set Membership: |
|

st14b
Genomic Map