Assay Details
| Gene Symbol: ydjc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082862.1 | NP_001076331.1 | 2 - 3 | 328 | 58 | |
| XM_005165050.4 | XP_005165107.1 | 3 - 4 | 608 | 58 | ||
More Information
Set Membership: |
|

ydjc
Genomic Map