Assay Details
| Gene Symbol: rab39ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098741.1 | NP_001092211.1 | 2 - 3 | 523 | 92 | |
| GenBank mRNA | BC142804.1 | - | 2 - 3 | 523 | 92 | |
More Information
Set Membership: |
|

rab39ba
Genomic Map