Assay Details
| Gene Symbol: lrp2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098776.1 | NP_001092246.1 | 3 - 4 | 541 | 86 | |
| XM_009307679.3 | XP_009305954.1 | 3 - 4 | 553 | 86 | ||
| GenBank mRNA | BC142915.1 | - | 3 - 4 | 541 | 86 | |
More Information
Set Membership: |
|

lrp2bp
Genomic Map