Assay Details
| Gene Symbol: rangrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099995.1 | NP_001093465.1 | 2 - 3 | 178 | 54 | |
| XM_005165494.4 | XP_005165551.1 | 3 - 4 | 556 | 54 | ||
| XM_005165495.2 | XP_005165552.1 | 3 - 4 | 257 | 54 | ||
| XM_009301822.3 | XP_009300097.1 | 3 - 4 | 358 | 54 | ||
| GenBank mRNA | BC095195.1 | - | 3 - 4 | 506 | 54 | |
| BC122253.1 | - | 3 - 4 | 513 | 54 | ||
| BC133096.1 | - | 2 - 3 | 626 | 54 | ||
More Information
Set Membership: |
|

rangrf
Genomic Map