Assay Details
| Gene Symbol: rangrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099995.1 | NP_001093465.1 | 4 - 5 | 443 | 98 | |
| XM_005165494.4 | XP_005165551.1 | 5 - 6 | 821 | 98 | ||
| XM_005165495.2 | XP_005165552.1 | 5 - 6 | 522 | 98 | ||
| XM_009301822.3 | XP_009300097.1 | 5 - 6 | 623 | 98 | ||
| GenBank mRNA | BC095195.1 | - | 5 - 6 | 771 | 98 | |
| BC122253.1 | - | 5 - 6 | 778 | 98 | ||
| BC150378.1 | - | 4 - 5 | 936 | 98 | ||
More Information
Set Membership: |
|

rangrf
Genomic Map