Assay Details
| Gene Symbol: oxnad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110456.1 | NP_001103926.1 | 6 - 7 | 752 | 68 | |
| XM_005170040.4 | XP_005170097.1 | 7 - 8 | 866 | 68 | ||
| XM_005170041.4 | XP_005170098.1 | 7 - 8 | 846 | 68 | ||
| GenBank mRNA | BC134194.1 | - | 6 - 7 | 752 | 68 | |
More Information
Set Membership: |
|

oxnad1
Genomic Map