Assay Details
| Gene Symbol: dbx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131158.1 | NP_571233.1 | 1 - 2 | 446 | 92 | |
| GenBank mRNA | AF030284.1 | - | 1 - 2 | 446 | 92 | |
| BC095020.1 | - | 1 - 2 | 452 | 92 | ||
| BC164106.1 | - | 1 - 2 | 386 | 92 | ||
More Information
Set Membership: |
|

dbx1a
Genomic Map