Assay Details
| Gene Symbol: hsp90ab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131310.3 | NP_571385.2 | 11 - 12 | 2183 | 66 | |
| GenBank mRNA | AF042108.1 | - | 11 - 12 | 2103 | 66 | |
| AF068772.1 | - | 11 - 12 | 2122 | 66 | ||
| BC065359.1 | - | 11 - 12 | 2129 | 66 | ||
More Information
Set Membership: |
|

hsp90ab1
Genomic Map