Assay Details
| Gene Symbol: gpia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144763.1 | NP_658909.1 | 16 - 17 | 1564 | 90 | |
| GenBank mRNA | AJ306395.1 | - | 16 - 17 | 1564 | 90 | |
| BC044450.1 | - | 16 - 17 | 1550 | 90 | ||
| BC083507.1 | - | 16 - 17 | 1548 | 90 | ||
| BC165764.1 | - | 16 - 17 | 1505 | 90 | ||
More Information
Set Membership: |
|

gpia
Genomic Map