Assay Details
| Gene Symbol: gpia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144763.1 | NP_658909.1 | 6 - 7 | 724 | 84 | |
| GenBank mRNA | AJ306395.1 | - | 6 - 7 | 724 | 84 | |
| BC044450.1 | - | 6 - 7 | 710 | 84 | ||
| BC083507.1 | - | 6 - 7 | 708 | 84 | ||
| BC165764.1 | - | 6 - 7 | 665 | 84 | ||
More Information
Set Membership: |
|

gpia
Genomic Map