Assay Details
| Gene Symbol: mapk14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313759.1 | NP_001300688.1 | 3 - 4 | 547 | 73 | |
| XM_017358195.2 | XP_017213684.1 | 3 - 4 | 575 | 73 | ||
| GenBank mRNA | AB030898.1 | - | 3 - 4 | 557 | 73 | |
| BC063937.1 | - | 3 - 4 | 505 | 73 | ||
| EH447521.1 | - | 3 - 4 | 558 | 73 | ||
More Information
Set Membership: |
|

mapk14b
Genomic Map