Assay Details
| Gene Symbol: ankhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194370.1 | NP_919351.1 | 1 - 2 | 358 | 70 | |
| GenBank mRNA | AJ302035.1 | - | 1 - 2 | 358 | 70 | |
| BC081575.1 | - | 1 - 2 | 419 | 70 | ||
More Information
Set Membership: |
|

ankhb
Genomic Map