Assay Details
| Gene Symbol: ankhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194370.1 | NP_919351.1 | 7 - 8 | 1181 | 119 | |
| GenBank mRNA | AJ302035.1 | - | 7 - 8 | 1181 | 119 | |
| BC081575.1 | - | 7 - 8 | 1242 | 119 | ||
More Information
Set Membership: |
|

ankhb
Genomic Map