Assay Details
| Gene Symbol: fhl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199217.1 | NP_954687.1 | 5 - 6 | 747 | 70 | |
| XM_009305310.3 | XP_009303585.1 | 5 - 6 | 824 | 70 | ||
| GenBank mRNA | BC053279.1 | - | 5 - 6 | 747 | 70 | |
| BC159222.1 | - | 5 - 6 | 770 | 70 | ||
More Information
Set Membership: |
|

fhl1b
Genomic Map