Assay Details
| Gene Symbol: alox12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199618.1 | NP_955912.1 | 3 - 4 | 358 | 70 | |
| GenBank mRNA | BC054621.1 | - | 3 - 4 | 358 | 70 | |
More Information
Set Membership: |
|

alox12
Genomic Map