Assay Details
| Gene Symbol: uck2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199764.2 | NP_956058.1 | 3 - 4 | 466 | 91 | |
| GenBank mRNA | BC045968.1 | - | 3 - 4 | 411 | 91 | |
| BC164836.1 | - | 3 - 4 | 402 | 91 | ||
| CB357915.1 | - | 3 - 4 | 401 | 91 | ||
More Information
Set Membership: |
|

uck2b
Genomic Map