Assay Details
| Gene Symbol: uck2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199764.2 | NP_956058.1 | 5 - 6 | 695 | 127 | |
| GenBank mRNA | BC045968.1 | - | 5 - 6 | 640 | 127 | |
| BC164836.1 | - | 5 - 6 | 631 | 127 | ||
| EH512314.1 | - | 2 - 3 | 471 | 127 | ||
More Information
Set Membership: |
|

uck2b
Genomic Map