Assay Details
| Gene Symbol: uck2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199764.2 | NP_956058.1 | 6 - 7 | 750 | 111 | |
| GenBank mRNA | BC045968.1 | - | 6 - 7 | 695 | 111 | |
| BC164836.1 | - | 6 - 7 | 686 | 111 | ||
| EH512314.1 | - | 1 - 2 | 416 | 111 | ||
More Information
Set Membership: |
|

uck2b
Genomic Map