Assay Details
| Gene Symbol: tk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199832.2 | NP_956126.1 | 2 - 3 | 187 | 116 | |
| GenBank mRNA | AY192987.1 | - | 2 - 3 | 100 | 116 | |
| BC044148.1 | - | 2 - 3 | 172 | 116 | ||
| BC164324.1 | - | 2 - 3 | 134 | 116 | ||
| EH580836.1 | - | 2 - 3 | 116 | |||
More Information
Set Membership: |
|

tk1
Genomic Map