Assay Details
| Gene Symbol: ap1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328014.1 | NP_001314943.1 | 3 - 4 | 454 | 252 | |
| NM_001328016.1 | NP_001314945.1 | 4 - 5 | 480 | 252 | ||
| NM_200309.2 | NP_956603.2 | 3 - 4 | 301 | 99 | ||
| GenBank mRNA | BC050180.1 | - | 3 - 4 | 228 | 99 | |
| BC070003.1 | - | 3 - 4 | 401 | 252 | ||
More Information
Set Membership: |
|

ap1s1
Genomic Map