Assay Details
| Gene Symbol: tmed5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200403.1 | NP_956697.1 | 2 - 3 | 297 | 102 | |
| GenBank mRNA | BC054575.1 | - | 2 - 3 | 297 | 102 | |
More Information
Set Membership: |
|

tmed5
Genomic Map