Assay Details
| Gene Symbol: wdr46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200623.1 | NP_956917.1 | 10 - 11 | 1189 | 56 | |
| GenBank mRNA | AY648746.1 | - | 10 - 11 | 1188 | 56 | |
| BC056828.1 | - | 10 - 11 | 1189 | 56 | ||
More Information
Set Membership: |
|

wdr46
Genomic Map