Assay Details
| Gene Symbol: prmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200650.2 | NP_956944.2 | 9 - 10 | 1053 | 67 | |
| GenBank mRNA | BC044522.1 | - | 9 - 10 | 1054 | 67 | |
| BC057480.1 | - | 9 - 10 | 1005 | 67 | ||
More Information
Set Membership: |
|

prmt1
Genomic Map