Assay Details
| Gene Symbol: med27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200660.1 | NP_956954.1 | 4 - 5 | 595 | 64 | |
| XM_021478216.1 | XP_021333891.1 | 4 - 5 | 615 | 64 | ||
| GenBank mRNA | BC057508.1 | - | 4 - 5 | 595 | 64 | |
| BC164095.1 | - | 4 - 5 | 608 | 64 | ||
More Information
Set Membership: |
|

med27
Genomic Map