Assay Details
| Gene Symbol: scp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200865.2 | NP_957159.1 | 13 - 14 | 1397 | 101 | |
| XM_005163230.2 | XP_005163287.2 | 6 - 7 | 709 | 101 | ||
| GenBank mRNA | BC063230.1 | - | 13 - 14 | 1331 | 101 | |
More Information
Set Membership: |
|

scp2a
Genomic Map