Assay Details
| Gene Symbol: scp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200865.2 | NP_957159.1 | 14 - 15 | 1528 | 67 | |
| XM_005163230.2 | XP_005163287.2 | 7 - 8 | 840 | 67 | ||
| GenBank mRNA | BC063230.1 | - | 14 - 15 | 1462 | 67 | |
More Information
Set Membership: |
|

scp2a
Genomic Map