Assay Details
| Gene Symbol: ahcyl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201340.1 | NP_958497.1 | 6 - 7 | 860 | 71 | |
| GenBank mRNA | BC059517.1 | - | 6 - 7 | 860 | 71 | |
More Information
Set Membership: |
|

ahcyl2b
Genomic Map