Assay Details
| Gene Symbol: smim14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205602.2 | NP_991165.1 | 4 - 5 | 422 | 88 | |
| GenBank mRNA | AY394928.1 | - | 4 - 5 | 461 | 88 | |
| BC068038.1 | - | 4 - 5 | 422 | 88 | ||
More Information
Set Membership: |
|

smim14
Genomic Map