Assay Details
| Gene Symbol: rbbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212595.1 | NP_997760.1 | 5 - 6 | 763 | 88 | |
| GenBank mRNA | AY394940.1 | - | 5 - 6 | 763 | 88 | |
| BC063984.1 | - | 5 - 6 | 676 | 88 | ||
More Information
Set Membership: |
|

rbbp4
Genomic Map