Assay Details
| Gene Symbol: gyg1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213510.1 | NP_998675.1 | 3 - 4 | 371 | 104 | |
| GenBank mRNA | BC044387.1 | - | 3 - 4 | 371 | 104 | |
| BC068329.1 | - | 3 - 4 | 357 | 104 | ||
| BC097080.1 | - | 3 - 4 | 371 | 104 | ||
| BC164360.1 | - | 2 - 3 | 344 | 104 | ||
| Gene Symbol: cep120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017357388.2 | XP_017212877.1 | 1046 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017357388.2 transcripts.
Set Membership: |
|

gyg1a
Genomic Map