Assay Details
| Gene Symbol: tnfsf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002593.2 | NP_001002593.1 | 1 - 2 | 189 | 97 | |
| NM_001328527.1 | NP_001315456.1 | 1 - 2 | 189 | 97 | ||
| GenBank mRNA | BC076005.1 | - | 1 - 2 | 140 | 97 | |
| EH448113.1 | - | 1 - 2 | 182 | 97 | ||
| EH499289.1 | - | 1 - 2 | 185 | 97 | ||
More Information
Set Membership: |
|

tnfsf10
Genomic Map