Assay Details
| Gene Symbol: sptlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003562.3 | NP_001003562.2 | 5 - 6 | 966 | 72 | |
| XM_021480563.1 | XP_021336238.1 | 5 - 6 | 1119 | 72 | ||
| XM_021480564.1 | XP_021336239.1 | 5 - 6 | 1119 | 72 | ||
| XM_021480565.1 | XP_021336240.1 | 5 - 6 | 1132 | 72 | ||
| GenBank mRNA | BC078234.1 | - | 5 - 6 | 952 | 72 | |
More Information
Set Membership: |
|

sptlc3
Genomic Map