Assay Details
| Gene Symbol: gpr34l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007217.1 | NP_001007218.1 | 1 | 1047 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gpr34l| Gene Symbol: gpr34l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007217.1 | NP_001007218.1 | 1 | 1047 | 117 | |
Set Membership: |
|