Assay Details
| Gene Symbol: slc27a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013537.1 | NP_001013555.1 | 10 - 11 | 1717 | 81 | |
| GenBank mRNA | BC090824.1 | - | 10 - 11 | 1717 | 81 | |
| BC165634.1 | - | 9 - 10 | 1511 | 81 | ||
More Information
Set Membership: |
|

slc27a1a
Genomic Map