Assay Details
| Gene Symbol: gabra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017822.1 | NP_001017822.1 | 4 - 5 | 850 | 56 | |
| GenBank mRNA | BC092794.1 | - | 4 - 5 | 850 | 56 | |
| BC164581.1 | - | 4 - 5 | 534 | 56 | ||
More Information
Set Membership: |
|

gabra4
Genomic Map