Assay Details
| Gene Symbol: ism2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020509.1 | NP_001018345.1 | 1 - 2 | 128 | 82 | |
| GenBank mRNA | AY856372.1 | - | 1 - 2 | 128 | 82 | |
| BC116497.1 | - | 1 - 2 | 520 | 82 | ||
More Information
Set Membership: |
|

ism2b
Genomic Map