Assay Details
| Gene Symbol: lats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020510.1 | NP_001018346.1 | 2 - 3 | 448 | 65 | |
| XM_005160312.4 | XP_005160369.1 | 3 - 4 | 696 | 65 | ||
| XM_009294504.3 | XP_009292779.1 | 3 - 4 | 699 | 65 | ||
| XM_021468417.1 | XP_021324092.1 | 3 - 4 | 699 | 65 | ||
| GenBank mRNA | AY957463.1 | - | 2 - 3 | 448 | 65 | |
| BC155305.1 | - | 3 - 4 | 566 | 65 | ||
| BC171344.1 | - | 2 - 3 | 501 | 65 | ||
More Information
Set Membership: |
|

lats1
Genomic Map