Assay Details
| Gene Symbol: miox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030266.1 | NP_001025437.1 | 4 - 5 | 360 | 77 | |
| GenBank mRNA | BC097218.1 | - | 4 - 5 | 360 | 77 | |
More Information
Set Membership: |
|

miox
Genomic Map