Assay Details
| Gene Symbol: pgghg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077725.1 | NP_001071193.1 | 9 - 10 | 1553 | 54 | |
| GenBank mRNA | BC125951.1 | - | 9 - 10 | 1553 | 54 | |
More Information
Set Membership: |
|

pgghg
Genomic Map