Assay Details
| Gene Symbol: mrps30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089371.1 | NP_001082840.1 | 2 - 3 | 726 | 77 | |
| GenBank mRNA | BC134231.1 | - | 2 - 3 | 726 | 77 | |
| BC152069.1 | - | 2 - 3 | 729 | 77 | ||
More Information
Set Membership: |
|

mrps30
Genomic Map