Assay Details
| Gene Symbol: zgc:162879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089427.1 | NP_001082896.1 | 7 - 8 | 1113 | 76 | |
| GenBank mRNA | BC139657.1 | - | 7 - 8 | 1113 | 76 | |
More Information
Set Membership: |
|

zgc:162879
Genomic Map