Assay Details
| Gene Symbol: mrpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100044.2 | NP_001093514.2 | 4 - 5 | 608 | 68 | |
| GenBank mRNA | BC153667.1 | - | 4 - 5 | 608 | 68 | |
More Information
Set Membership: |
|

mrpl9
Genomic Map