Assay Details
| Gene Symbol: trioa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104526.1 | NP_001097996.1 | 10 - 11 | 1879 | 67 | |
| XM_009294421.3 | XP_009292696.1 | 10 - 11 | 2046 | 67 | ||
| XM_017352243.2 | XP_017207732.1 | 10 - 11 | 2050 | 67 | ||
| XM_021468075.1 | XP_021323750.1 | 10 - 11 | 2109 | 67 | ||
| XM_021468076.1 | XP_021323751.1 | 10 - 11 | 2110 | 67 | ||
| XM_021468077.1 | XP_021323752.1 | 10 - 11 | 2110 | 67 | ||
| XM_021468078.1 | XP_021323753.1 | 10 - 11 | 2111 | 67 | ||
| XM_021468079.1 | XP_021323754.1 | 10 - 11 | 2112 | 67 | ||
| XM_021468080.1 | XP_021323755.1 | 10 - 11 | 2113 | 67 | ||
| XM_021468081.1 | XP_021323756.1 | 10 - 11 | 2113 | 67 | ||
More Information
Set Membership: |
|

trioa
Genomic Map