Assay Details
| Gene Symbol: trioa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104526.1 | NP_001097996.1 | 2 - 3 | 232 | 68 | |
| XM_009294421.3 | XP_009292696.1 | 2 - 3 | 399 | 68 | ||
| XM_017352243.2 | XP_017207732.1 | 2 - 3 | 403 | 68 | ||
| XM_021468075.1 | XP_021323750.1 | 2 - 3 | 399 | 68 | ||
| XM_021468076.1 | XP_021323751.1 | 2 - 3 | 400 | 68 | ||
| XM_021468077.1 | XP_021323752.1 | 2 - 3 | 400 | 68 | ||
| XM_021468078.1 | XP_021323753.1 | 2 - 3 | 401 | 68 | ||
| XM_021468079.1 | XP_021323754.1 | 2 - 3 | 402 | 68 | ||
| XM_021468080.1 | XP_021323755.1 | 2 - 3 | 403 | 68 | ||
| XM_021468081.1 | XP_021323756.1 | 2 - 3 | 403 | 68 | ||
More Information
Set Membership: |
|

trioa
Genomic Map