Assay Details
| Gene Symbol: elavl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308614.1 | NP_001295543.1 | 6 - 7 | 659 | 85 | |
| NM_001308615.1 | NP_001295544.1 | 5 - 6 | 599 | 85 | ||
| NM_001308616.1 | NP_001295545.1 | 5 - 6 | 586 | 85 | ||
| NM_001308617.1 | NP_001295546.1 | 6 - 7 | 649 | 85 | ||
| NM_001308618.1 | NP_001295547.1 | 4 - 5 | 495 | 85 | ||
| NM_001308619.1 | NP_001295548.1 | 5 - 6 | 558 | 85 | ||
| NM_130909.2 | NP_570984.2 | 6 - 7 | 662 | 85 | ||
| XM_017358196.2 | XP_017213685.1 | 5 - 6 | 660 | 85 | ||
| XM_017358197.2 | XP_017213686.1 | 4 - 5 | 508 | 85 | ||
| GenBank mRNA | AF184245.1 | - | 6 - 7 | 661 | 85 | |
| BC045277.1 | - | 4 - 5 | 432 | 85 | ||
| BC165930.1 | - | 1 - 2 | 116 | 85 | ||
More Information
Set Membership: |
|

elavl1b
Genomic Map